5-Bromo-4-phenyl-1,3-thiazol-2-amine hydrobromide ≥95%
Supplier: Matrix Scientific
048171-500MG
048171-1G
048171-5G
101883-144EA
264.62
USD
101883-144
101883-146
101883-148
5-Bromo-4-phenyl-1,3-thiazol-2-amine hydrobromide ≥95%
5-Bromo-4-phenyl-1,3-thiazol-2-amine hydrobromide
Hazard: Irritant
Formula:
C₉H₈Br₂N₂S Melting Pt: 195…197 °C |
MDL Number:
MFCD09972265 CAS Number: 113511-22-1 |
Specification Test Results
Melting point | 195-197°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...