N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine ≥95%

Supplier: Matrix Scientific

048169-500MG 048169-1G 048169-5G
101883-132EA 264.62 USD
101883-132 101883-134 101883-136
N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine ≥95%
N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
Melting Pt: 125…126 °C
MDL Number: MFCD09972264
CAS Number: 14703-82-3

Order Now

Specification Test Results

Melting point 125-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR