3-Methoxy-N-methyl-4-nitroaniline ≥95%

Supplier: Matrix Scientific

048168-500MG 048168-1G 048168-5G
101883-126EA 264.62 USD
101883-126 101883-128 101883-130
3-Methoxy-N-methyl-4-nitroaniline ≥95%
3-Methoxy-N-methyl-4-nitroaniline

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
Melting Pt: 126…127 °C
MDL Number: MFCD09972263
CAS Number: 1017782-97-6

Order Now

Specification Test Results

Melting point 126-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR