1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-(4-Chlorophenyl)thiazol-2-yl)piperidine-4-carboxylic acid
048167-500MG
048167-1G
101883-122EA
375.38
USD
101883-122
101883-124
1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid ≥95%
1-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₅ClN₂O₂S MW: 322.82 g/mol Melting Pt: 187…189 °C |
MDL Number:
MFCD01899691 CAS Number: 345990-26-3 |
Specification Test Results
Melting point | 187-189°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...