1-(3-Methoxy-4-nitrophenyl)piperazine ≥95%
Supplier: Matrix Scientific
048164-500MG
048164-1G
048164-5G
101883-110EA
264.62
USD
101883-110
101883-112
101883-114
1-(3-Methoxy-4-nitrophenyl)piperazine ≥95%
1-(3-Methoxy-4-nitrophenyl)piperazine
Hazard: Irritant
Formula:
C₁₁H₁₅N₃O₃ Melting Pt: 140…142 °C |
MDL Number:
MFCD00546013 CAS Number: 121278-37-3 |
Specification Test Results
Melting point | 140-142°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...