(6-Morpholino-3-pyridinyl)-1-ethanone ≥97%
Supplier: Matrix Scientific
048163-500MG
048163-1G
101883-106EA
221.54
USD
101883-106
101883-108
(6-Morpholino-3-pyridinyl)-1-ethanone ≥97%
(6-Morpholino-3-pyridinyl)-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ Melting Pt: 78-80°C |
MDL Number:
MFCD09910348 CAS Number: 265107-43-5 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...