3-(Dimethylamino)-1-[4-(2-pyrimidinyloxy)phenyl]-2-buten-1-one ≥95%
Supplier: Matrix Scientific
048161-500MG
048161-1G
101883-100EA
375.38
USD
101883-100
101883-102
3-(Dimethylamino)-1-[4-(2-pyrimidinyloxy)phenyl]-2-buten-1-one ≥95%
3-(Dimethylamino)-1-[4-(2-pyrimidinyloxy)phenyl]-2-buten-1-one
Hazard: Irritant
Formula:
C₁₆H₁₇N₃O₂ Melting Pt: 182…184 °C |
MDL Number:
MFCD09152761 CAS Number: 952182-85-3 |
Specification Test Results
Melting point | 182-184°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...