1,3-Benzothiazole-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

048160-500MG 048160-1G 048160-5G
101883-094EA 173.85 USD
101883-094 101883-096 101883-098
1,3-Benzothiazole-2-carbohydrazide ≥95%
1,3-Benzothiazole-2-carbohydrazide

Hazard: Irritant

Formula: C₈H₇N₃OS
Melting Pt: 177…179 °C
MDL Number: MFCD02713577
CAS Number: 28891-34-1

Order Now

Specification Test Results

Melting point 177-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR