Ethyl-3-[(5-bromo-2-pyrimidinyl)oxy]-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
048154-500MG
048154-1G
101883-080EA
921.54
USD
101883-080
101883-082
Ethyl-3-[(5-bromo-2-pyrimidinyl)oxy]-2-thiophenecarboxylate ≥95%
Ethyl-3-[(5-bromo-2-pyrimidinyl)oxy]-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₉BrN₂O₃S Melting Pt: 120…122 °C |
MDL Number:
MFCD09152759 CAS Number: 952182-83-1 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...