Methyl-2-chloro-4-piperidinobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
048150-500MG
048150-1G
048150-5G
101883-056EA
278.46
USD
101883-056
101883-058
101883-060
Methyl-2-chloro-4-piperidinobenzenecarboxylate ≥95%
Methyl-2-chloro-4-piperidinobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₆ClNO₂ Melting Pt: 37 °C |
MDL Number:
MFCD09152756 CAS Number: 313674-08-7 |
Specification Test Results
Melting point | 37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...