Ethyl-4-(2-naphthylcarbonyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

048149-500MG 048149-1G 048149-5G
101883-050EA 375.38 USD
101883-050 101883-052 101883-054
Ethyl-4-(2-naphthylcarbonyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(2-naphthylcarbonyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₇H₁₃NO₄
Melting Pt: 97…99 °C
MDL Number: MFCD09152755
CAS Number: 952182-79-5

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR