Ethyl-4-(4-chlorobenzoyl)-3-isoxazolecarboxylate ≥95%
Supplier: Matrix Scientific
048148-500MG
048148-1G
048148-5G
101883-044EA
356.92
USD
101883-044
101883-046
101883-048
Ethyl-4-(4-chlorobenzoyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(4-chlorobenzoyl)-3-isoxazolecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₀ClNO₄ Melting Pt: 86…88 °C |
MDL Number:
MFCD09152754 CAS Number: 952182-77-3 |
Specification Test Results
Melting point | 86-88°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...