2-Chloro-4-(1-pyrrolidinyl)benzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
048147-500MG
048147-1G
048147-5G
101883-038EA
375.38
USD
101883-038
101883-040
101883-042
2-Chloro-4-(1-pyrrolidinyl)benzenecarbohydrazide ≥95%
2-Chloro-4-(1-pyrrolidinyl)benzenecarbohydrazide
Hazard: Irritant
Formula:
C₁₁H₁₄ClN₃O Melting Pt: 127…129 °C |
MDL Number:
MFCD09152753 CAS Number: 952182-75-1 |
Specification Test Results
Melting point | 127-129°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...