3-Mercaptobenzoic acid ≥94%

Supplier: Matrix Scientific

048146-1G 048146-5G
101883-034EA 76.92 USD
101883-034 101883-036
3-Mercaptobenzoic acid ≥94%
3-Mercaptobenzoic acid

Hazard: Irritant

Formula: C₇H₆O₂S
MW: 154.1863 g/mol
Melting Pt: 144-147°C
MDL Number: MFCD00238636
CAS Number: 4869-59-4

Order Now

Specification Test Results

Melting point 144-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR