5,11-Dihydro-6H-indolo[3,2-c]quinolin-6-one ≥95%

Supplier: Matrix Scientific

048144-500MG 048144-1G 048144-5G
101883-026EA 264.62 USD
101883-026 101883-028 101883-030
5,11-Dihydro-6H-indolo[3,2-c]quinolin-6-one ≥95%
5,11-Dihydro-6H-indolo[3,2-c]quinolin-6-one

Hazard: Irritant

Formula: C₁₅H₁₀N₂O
Melting Pt: 379…381 °C
MDL Number: MFCD00225019
CAS Number: 18735-98-3

Order Now

Specification Test Results

Melting point 379-381°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR