4-[(4-Bromophenyl)thio]piperidine hydrochloride ≥95%
Supplier: Matrix Scientific
048143-500MG
048143-1G
101883-022EA
747.69
USD
101883-022
101883-024
4-[(4-Bromophenyl)thio]piperidine hydrochloride ≥95%
4-[(4-Bromophenyl)thio]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅BrClNS Melting Pt: 175…177 °C |
MDL Number:
MFCD09152752 CAS Number: 1134681-62-1 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...