2,4,6-Trichlorophenyl-2-phenyl-1-ethylenesulfonate ≥95%

Supplier: Matrix Scientific

048141-500MG 048141-1G 048141-5G
101883-016EA 278.46 USD
101883-016 101883-018 101883-020
2,4,6-Trichlorophenyl-2-phenyl-1-ethylenesulfonate ≥95%
2,4,6-Trichlorophenyl-2-phenyl-1-ethylenesulfonate

Hazard: Irritant

Formula: C₁₄H₉Cl₃O₃S
Melting Pt: 141…142 °C
MDL Number: MFCD00988346
CAS Number: 325857-42-9

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR