2,4,6-Trichlorophenyl-1-propanesulfonate ≥95%

Supplier: Matrix Scientific

048140-500MG 048140-1G 048140-5G
101883-010EA 278.46 USD
101883-010 101883-012 101883-014
2,4,6-Trichlorophenyl-1-propanesulfonate ≥95%
2,4,6-Trichlorophenyl 1-propanesulphonate

Hazard: Irritant

Formula: C₉H₉Cl₃O₃S
Melting Pt: 71-73°C
MDL Number: MFCD12498735
CAS Number: 1171919-51-9

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR