2,4,6-Trichlorophenyl-2-naphthalenesulfonate ≥95%

Supplier: Matrix Scientific

048136-500MG 048136-1G 048136-5G
101882-986EA 278.46 USD
101882-986 101882-988 101882-990
2,4,6-Trichlorophenyl-2-naphthalenesulfonate ≥95%
2,4,6-Trichlorophenyl-2-naphthalenesulfonate

Hazard: Irritant

Formula: C₁₆H₉Cl₃O₃S
Melting Pt: 128…130 °C
MDL Number: MFCD12498731
CAS Number: 1171919-42-8

Order Now

Specification Test Results

Melting point 128-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR