2,4,6-Trichlorophenyl-4-fluorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

048134-500MG 048134-1G 048134-5G
101882-974EA 278.46 USD
101882-974 101882-976 101882-978
2,4,6-Trichlorophenyl-4-fluorobenzenesulfonate ≥95%
2,4,6-Trichlorophenyl 4-fluorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₆Cl₃FO₃S
Melting Pt: 114-116°C
MDL Number: MFCD12498729
CAS Number: 1171919-37-1

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR