2,4,6-Trichlorophenyl-2-fluorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

048133-500MG 048133-1G 048133-5G
101882-968EA 278.46 USD
101882-968 101882-970 101882-972
2,4,6-Trichlorophenyl-2-fluorobenzenesulfonate ≥95%
2,4,6-Trichlorophenyl 2-fluorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₆Cl₃FO₃S
Melting Pt: 91-93°C
MDL Number: MFCD12498728
CAS Number: 1171919-36-0

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR