2,4,6-Trichlorophenyl phenylmethanesulfonate ≥95%

Supplier: Matrix Scientific

048131-500MG 048131-1G 048131-5G
101882-956EA 278.46 USD
101882-956 101882-958 101882-960
2,4,6-Trichlorophenyl phenylmethanesulfonate ≥95%
2,4,6-Trichlorophenyl phenylmethanesulfonate

Hazard: Irritant

Formula: C₁₃H₉Cl₃O₃S
MW: 351.64 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD01231356
CAS Number: 1171919-33-7

Order Now

Specification Test Results

Melting point 107-109°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR