2,4,6-Trichlorophenyl-2-cyanobenzenesulfonate ≥95%

Supplier: Matrix Scientific

048129-500MG 048129-1G 048129-5G
101882-944EA 278.46 USD
101882-944 101882-946 101882-948
2,4,6-Trichlorophenyl-2-cyanobenzenesulfonate ≥95%
2,4,6-Trichlorophenyl-2-cyanobenzenesulfonate

Hazard: Irritant

Formula: C₁₃H₆Cl₃NO₃S
Melting Pt: 178…181 °C
MDL Number: MFCD12498725
CAS Number: 1171919-29-1

Order Now

Specification Test Results

Melting point 178-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR