2,4,6-Trichlorophenyl-4-(trifluoromethoxy)benzenesulfonate ≥95%

Supplier: Matrix Scientific

048128-500MG 048128-1G 048128-5G
101882-938EA 278.46 USD
101882-938 101882-940 101882-942
2,4,6-Trichlorophenyl-4-(trifluoromethoxy)benzenesulfonate ≥95%
2,4,6-Trichlorophenyl 4-(trifluoromethoxy)benzenesulphonate

Hazard: Irritant

Formula: C₁₃H₆Cl₃F₃O₄S
Melting Pt: 72-74°C
MDL Number: MFCD12498724
CAS Number: 1171919-27-9

Order Now

Specification Test Results

Melting point 72-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR