2,4,6-Trichlorophenyl-4-methylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

048126-500MG 048126-1G 048126-5G
101882-926EA 278.46 USD
101882-926 101882-928 101882-930
2,4,6-Trichlorophenyl-4-methylbenzenesulfonate ≥95%
2,4,6-Trichlorophenyl-4-methylbenzenesulfonate

Hazard: Irritant

Formula: C₁₃H₉Cl₃O₃S
Melting Pt: 84…87 °C
MDL Number: MFCD00422258
CAS Number: 7466-04-8

Order Now

Specification Test Results

Melting point 82-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR