2,4,6-Trichlorophenyl-4-bromobenzenesulfonate ≥95%
Supplier: Matrix Scientific
048125-500MG
048125-1G
048125-5G
101882-920EA
278.46
USD
101882-920
101882-922
101882-924
2,4,6-Trichlorophenyl-4-bromobenzenesulfonate ≥95%
2,4,6-Trichlorophenyl-4-bromobenzenesulfonate
Hazard: Irritant
Formula:
C₁₂H₆BrCl₃O₃S Melting Pt: 112…114 °C |
MDL Number:
MFCD12498722 CAS Number: 934986-62-6 |
Specification Test Results
Melting point | 112-114°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...