2,4,6-Trichlorophenyl-8-quinolinesulfonate ≥95%
Supplier: Matrix Scientific
048124-500MG
048124-1G
048124-5G
101882-914EA
278.46
USD
101882-914
101882-916
101882-918
2,4,6-Trichlorophenyl-8-quinolinesulfonate ≥95%
2,4,6-Trichlorophenyl-8-quinolinesulfonate
Hazard: Irritant
Formula:
C₁₅H₈Cl₃NO₃S Melting Pt: 160…162 °C |
MDL Number:
MFCD12498721 CAS Number: 1171919-22-4 |
Specification Test Results
Melting point | 160-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...