2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥95%

Supplier: Matrix Scientific

048119-5G 048119-25G
101882-888EA 30.77 USD
101882-888 101882-890
2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥95%
2,2,3,3-Tetramethylcyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
Melting Pt: 120…123 °C
MDL Number: MFCD00011538
CAS Number: 15641-58-4

Order Now

Specification Test Results

Melting point 120-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR