3-Benzyl-3-azabicyclo[3.1.0]hexane-2,4-dione ≥95%

Supplier: Matrix Scientific

048118-500MG 048118-1G 048118-5G
101882-880EA 347.69 USD
101882-880 101882-882 101882-884
3-Benzyl-3-azabicyclo[3.1.0]hexane-2,4-dione ≥95%
3-Benzyl-3-azabicyclo[3.1.0]hexane-2,4-dione

Hazard: Irritant

Formula: C₁₂H₁₁NO₂
Melting Pt: 92…95 °C
MDL Number: MFCD00158669
CAS Number: 73799-63-0

Order Now

Specification Test Results

Melting point 92-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR