Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate ≥95%

Supplier: Matrix Scientific

048117-500MG 048117-1G 048117-5G
101882-874EA 278.46 USD
101882-874 101882-876 101882-878
Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate ≥95%
Ethyl-2-(benzoyl-3-chloro-4-fluoroanilino)propanoate

Hazard: Irritant

Formula: C₁₈H₁₇ClFNO₃
MW: 349.79 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD01310648
CAS Number: 34029-27-1

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR