Ethyl-2-(acetylamino)-4-(methylsulfanyl)butanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(acetylamino)-4-(methylthio)butanoate

048116-500MG 048116-1G 048116-5G
101882-868EA 375.38 USD
101882-868 101882-870 101882-872
Ethyl-2-(acetylamino)-4-(methylsulfanyl)butanoate ≥95%
Ethyl-2-(acetylamino)-4-(methylsulfanyl)butanoate

Hazard: Irritant

Formula: C₉H₁₇NO₃S
MW: 219.3 g/mol
Melting Pt: 60…62 °C
MDL Number: MFCD05663833
CAS Number: 33280-93-2

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR