4-Isopropyl-3-methylaniline hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methyl-4-(prop-2-yl)aniline hydrochloride, 3-Methyl-4-isopropylaniline hydrochloride, 4-Isopropyl-m-toluidinium chloride
048115-500MG
048115-1G
101882-862EA
181.54
USD
101882-862
101882-864
4-Isopropyl-3-methylaniline hydrochloride ≥95%
4-Isopropyl-3-methylaniline hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₆ClN MW: 185.70 g/mol Melting Pt: 213°C |
MDL Number:
MFCD00156562 CAS Number: 4534-11-6 |
Specification Test Results
Melting point | 224-227°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...