Methyl-6-chloropicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate

048113-1G 048113-5G 048113-25G
101882-856EA 15.38 USD
101882-856 101882-858 101882-860
Methyl-6-chloropicolinate ≥95%
Methyl 6-chloropicolinate

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 93…94 °C
MDL Number: MFCD00155391
CAS Number: 6636-55-1

Order Now

Specification Test Results

Melting point 93-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR