3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopropane-1,2-dicarboxylic acid anhydride

048111-1G 048111-5G
101882-848EA 26.15 USD
101882-848 101882-850
3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥95%
3-Oxabicyclo[3.1.0]hexane-2,4-dione

Hazard: Irritant

Formula: C5H4O3
MW: 112.08 g/mol
Boiling Pt: 100…102 °C (5 mmHg)
Melting Pt: 59…61 °C
Density: 1.5
MDL Number: MFCD00126929
CAS Number: 5617-74-3

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR