1-(2-Pyridinyl)-1-ethanone oxime ≥95%

Supplier: Matrix Scientific

048108-500MG 048108-1G
101882-832EA 375.38 USD
101882-832 101882-834
1-(2-Pyridinyl)-1-ethanone oxime ≥95%
1-(2-Pyridinyl)-1-ethanone oxime

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD00499500
CAS Number: 1758-54-9

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR