3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (1R,5S)-3-azabicyclo[3.1.0]hexane-4-carboxylic acid

048105-500MG 048105-1G 048105-5G
101882-816EA 615.38 USD
101882-816 101882-818 101882-820
3-Azabicyclo[3.1.0]hexane-2-carboxylic acid ≥95%
3-Azabicyclo[3.1.0]hexane-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₉NO₂
MW: 127.143 g/mol
Melting Pt: 252…256 °C
MDL Number: MFCD07021308
CAS Number: 22255-16-9

Order Now

Specification Test Results

Melting point 252-256°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR