1-[2-(Chloromethyl)-1,3-benzoxazol-7-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

048097-500MG 048097-1G
101882-782EA 278.46 USD
101882-782 101882-784
1-[2-(Chloromethyl)-1,3-benzoxazol-7-yl]-1-ethanone ≥95%
1-[2-(Chloromethyl)-1,3-benzoxazol-7-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₀H₈ClNO₂
Melting Pt: 77…78 °C
MDL Number: MFCD11841033
CAS Number: 1135283-66-7

Order Now

Specification Test Results

Melting point 77-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR