[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol ≥95%

Supplier: Matrix Scientific

048092-500MG 048092-1G
101882-764EA 232.31 USD
101882-764 101882-766
[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol ≥95%
[5-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol

Hazard: Irritant

Formula: C₁₀H₇F₃OS
Melting Pt: 105…107 °C
MDL Number: MFCD11841028
CAS Number: 951122-90-0

Order Now

Specification Test Results

Melting point 105-107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR