5-Methoxy-2-(4-pyridinyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

048089-500MG 048089-1G
101882-754EA 556.92 USD
101882-754 101882-756
5-Methoxy-2-(4-pyridinyl)-4-pyrimidinol ≥95%
5-Methoxy-2-(4-pyridinyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
Melting Pt: 248 °C (dec.)
MDL Number: MFCD11841025
CAS Number: 133661-37-7

Order Now

Specification Test Results

Melting point 248°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR