tert-Butyl-2-(4-aminobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

048088-500MG 048088-1G
101882-750EA 1109.23 USD
101882-750 101882-752
tert-Butyl-2-(4-aminobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%
tert-Butyl-2-(4-aminobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₃
MW: 305.38 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD11841024
CAS Number: 1135283-60-1

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR