tert-Butyl-2-(4-nitrobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%
Supplier: Matrix Scientific
048087-500MG
048087-1G
048087-5G
101882-744EA
921.54
USD
101882-744
101882-746
101882-748
tert-Butyl-2-(4-nitrobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate ≥95%
tert-Butyl-2-(4-nitrobenzoyl)tetrahydro-1(2H)-pyridazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁N₃O₅ MW: 335.36 g/mol Melting Pt: 106…108 °C |
MDL Number:
MFCD11553076 CAS Number: 443295-21-4 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...