3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Thiazolidine-2-carboxylic acid, N-BOC protected

048081-500MG 048081-1G 048081-5G
101882-728EA 278.46 USD
101882-728 101882-730 101882-732
3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid ≥95%
3-(tert-Butoxycarbonyl)-1,3-thiazolane-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄S
Melting Pt: 95 °C
MDL Number: MFCD01569306
CAS Number: 141783-63-3

Order Now

Specification Test Results

Melting point 95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR