4,4-Dimethyl-2-pyrrolidinone ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4-Dimethylpyrrolidin-2-one

048080-500MG 048080-1G
101882-724EA 40 USD
101882-724 101882-726
4,4-Dimethyl-2-pyrrolidinone ≥97%
4,4-Dimethyl-2-pyrrolidinone

Hazard: Irritant

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD09864496
CAS Number: 66899-02-3

Order Now

Specification Test Results

Melting point 38-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR