2-(4-Methylpiperazino)-5-nitrobenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
048079-500MG
048079-1G
048079-5G
101882-718EA
181.54
USD
101882-718
101882-720
101882-722
2-(4-Methylpiperazino)-5-nitrobenzenecarbaldehyde ≥95%
2-(4-Methylpiperazino)-5-nitrobenzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₅N₃O₃ Melting Pt: 150…153 °C |
MDL Number:
MFCD00431681 CAS Number: 30742-63-3 |
Specification Test Results
Melting point | 150-153°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...