(5-Bromo-3-pyridinyl)(piperazino)methanone ≥95%

Supplier: Matrix Scientific

048076-500MG 048076-1G
101882-702EA 269.23 USD
101882-702 101882-704
(5-Bromo-3-pyridinyl)(piperazino)methanone ≥95%
(5-Bromo-3-pyridinyl)(piperazino)methanone

Hazard: Irritant

Formula: C₁₀H₁₂BrN₃O
Melting Pt: 79…80 °C
MDL Number: MFCD09972258
CAS Number: 1017782-91-0

Order Now

Specification Test Results

Melting point 79-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR