2-(2-Cyanophenoxy)acetic acid ≥95%
Supplier: Matrix Scientific
048074-500MG
048074-1G
101882-694EA
76.92
USD
101882-694
101882-696
2-(2-Cyanophenoxy)acetic acid ≥95%
2-(2-Cyanophenoxy)acetic acid
Hazard: Irritant
Formula:
C₉H₇NO₃ Melting Pt: 181…182 °C |
MDL Number:
MFCD02257373 CAS Number: 6574-95-4 |
Specification Test Results
Melting point | 181-182°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...