2-(4-Methylphenyl)-1,3-thiazolane ≥95%

Supplier: Matrix Scientific

048072-500MG 048072-1G 048072-5G
101882-684EA 172.31 USD
101882-684 101882-686 101882-688
2-(4-Methylphenyl)-1,3-thiazolane ≥95%
2-(4-Methylphenyl)-1,3-thiazolane

Hazard: Irritant

Formula: C₁₀H₁₃NS
Melting Pt: 96-98°C
MDL Number: MFCD00222822
CAS Number: 67086-82-2

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR