Ethyl-1-(3-methoxy-4-nitrophenyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

048068-500MG 048068-1G 048068-5G
101882-662EA 278.46 USD
101882-662 101882-664 101882-666
Ethyl-1-(3-methoxy-4-nitrophenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(3-methoxy-4-nitrophenyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₅
Melting Pt: 113…115 °C
MDL Number: MFCD09972253
CAS Number: 1017782-81-8

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR