2-[4-(1,3-Benzothiazol-2-yl)piperazino]-1-ethanol ≥95%

Supplier: Matrix Scientific

048066-500MG 048066-1G 048066-5G
101882-650EA 278.46 USD
101882-650 101882-652 101882-654
2-[4-(1,3-Benzothiazol-2-yl)piperazino]-1-ethanol ≥95%
2-[4-(1,3-Benzothiazol-2-yl)piperazino]-1-ethanol

Hazard: Irritant

Formula: C₁₃H₁₇N₃OS
MW: 263.36 g/mol
Melting Pt: 103…104 °C
MDL Number: MFCD09382882
CAS Number: 232263-28-4

Order Now

Specification Test Results

Melting point 103-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR