2-{4-[(4-Methoxyphenyl)sulfonyl]piperazino}-1-ethanol ≥95%

Supplier: Matrix Scientific

048063-500MG 048063-1G 048063-5G
101882-638EA 106.15 USD
101882-638 101882-640 101882-642
2-{4-[(4-Methoxyphenyl)sulfonyl]piperazino}-1-ethanol ≥95%
2-{4-[(4-Methoxyphenyl)sulfonyl]piperazino}-1-ethanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₄S
Melting Pt: 121…123 °C
MDL Number: MFCD01211979
CAS Number: 321531-40-2

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR